Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROBO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002941.3 | NP_002932.1 | 2 | 274 | ||
XM_011533976.1 | XP_011532278.1 | 2 | 363 | |||
XM_011533977.2 | XP_011532279.1 | 2 | 216 | |||
XM_011533978.1 | XP_011532280.1 | 2 | 363 | |||
XM_011533979.1 | XP_011532281.1 | 2 | 363 | |||
XM_011533980.1 | XP_011532282.1 | 2 | 363 | |||
XM_017006982.1 | XP_016862471.1 | 2 | 359 | |||
XM_017006984.1 | XP_016862473.1 | 2 | 363 | |||
GenBank mRNA | AF040990.1 | - | 1 | 66 | ||
AK297481.1 | - | 1 | 222 | |||
DR003097.1 | - | 281 |