Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIGD1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109619.1 | NP_001103089.1 | 2 | 132 | ||
XM_011538649.2 | XP_011536951.1 | 4 | 279 | |||
XM_017019783.1 | XP_016875272.1 | 5 | 1707 | |||
XM_017019784.1 | XP_016875273.1 | 5 | 1707 | |||
XM_017019785.1 | XP_016875274.1 | 3 | 211 | |||
GenBank mRNA | DW009649.1 | - | 2 | 69 |