Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000999.3 | NP_000990.1 | 4 | 225 | ||
NM_001035258.1 | NP_001030335.1 | 4 | 190 | |||
GenBank mRNA | AK312119.1 | - | 4 | 193 | ||
BC000603.2 | - | 4 | 162 | 3346447 | ||
BQ276548.1 | - | 4 | 221 | |||
BU532782.1 | - | 4 | 193 | |||
CB124619.1 | - | 4 | 215 | |||
CB128529.1 | - | 4 | 168 | |||
Z26876.1 | - | 4 | 207 |