Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XYLT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022167.3 | NP_071450.2 | 9, 10 | 2048 | ||
XM_005257572.4 | XP_005257629.1 | 9, 10 | 2362 | |||
GenBank mRNA | AJ277442.1 | - | 9, 10 | 2048 | ||
BC052262.1 | - | 9, 10 | 1968 | 6380013 |