Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102469.1 | NP_001095939.1 | 1 | 57 | ||
XM_005270049.3 | XP_005270106.1 | 3 | 349 | |||
XM_011540083.2 | XP_011538385.1 | 3 | 386 | |||
XM_011540084.2 | XP_011538386.1 | 2 | 271 | |||
XM_011540085.2 | XP_011538387.1 | 3 | 386 | |||
XM_011540086.2 | XP_011538388.1 | 3 | 386 | |||
XM_017016549.1 | XP_016872038.1 | 3 | 386 | |||
GenBank mRNA | EF426483.1 | - | 1 | 57 |