Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A4E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011545417.2 | XP_011543719.1 | 4, 5 | 496 | ||
XM_011545418.2 | XP_011543720.1 | 3, 4 | 462 | |||
XM_011545419.2 | XP_011543721.1 | 3, 4 | 462 | |||
GenBank mRNA | AB231726.1 | - | 2, 3 | 274 | ||
AB231729.1 | - | 3, 4 | 302 | |||
AB231730.1 | - | 3, 4 | 308 | |||
AB231731.1 | - | 3, 4 | 308 | |||
AK300102.1 | - | 4, 5 | 460 |