Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130782.2 | NP_570138.1 | 5 | 795 | ||
XM_011509891.2 | XP_011508193.1 | 3 | 362 | |||
GenBank mRNA | AF076642.1 | - | 5 | 769 | ||
AF268036.1 | - | 5 | 781 | |||
AK057114.1 | - | 5 | 788 | |||
AK292015.1 | - | 5 | 787 | |||
AK315377.1 | - | 5 | 788 | |||
BC020632.1 | - | 5 | 787 | 4714909 |