Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STARD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020759.2 | NP_065810.2 | 3 | 224 | ||
XM_011521831.2 | XP_011520133.1 | 3 | 237 | |||
XM_011521832.2 | XP_011520134.1 | 3 | 237 | |||
XM_011521833.2 | XP_011520135.1 | 3 | 237 | |||
XM_011521834.2 | XP_011520136.1 | 3 | 237 | |||
XM_011521835.2 | XP_011520137.1 | 3 | 237 | |||
XM_011521836.2 | XP_011520138.1 | 3 | 539 | |||
XM_017022439.1 | XP_016877928.1 | 3 | 237 | |||
XM_017022440.1 | XP_016877929.1 | 2 | 136 | |||
GenBank mRNA | CR627426.1 | - | 3 | 224 |