Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STARD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020759.2 | NP_065810.2 | 11 | 895 | ||
XM_011521831.2 | XP_011520133.1 | 11 | 908 | |||
XM_011521832.2 | XP_011520134.1 | 11 | 905 | |||
XM_011521833.2 | XP_011520135.1 | 10 | 875 | |||
XM_011521834.2 | XP_011520136.1 | 11 | 908 | |||
XM_011521835.2 | XP_011520137.1 | 11 | 908 | |||
XM_011521836.2 | XP_011520138.1 | 11 | 1210 | |||
XM_011521837.2 | XP_011520139.1 | 9 | 677 | |||
XM_017022439.1 | XP_016877928.1 | 11 | 908 | |||
XM_017022440.1 | XP_016877929.1 | 10 | 807 | |||
XM_017022441.1 | XP_016877930.1 | 9 | 1126 | |||
XM_017022442.1 | XP_016877931.1 | 4 | 234 | |||
XM_017022443.1 | XP_016877932.1 | 3 | 218 | |||
GenBank mRNA | AK122666.1 | - | 9 | 909 | ||
CR627426.1 | - | 11 | 895 |