Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198978.1 | NP_001185907.1 | 7 | 610 | ||
NM_001198979.1 | NP_001185908.1 | 7 | 694 | |||
NM_001198980.1 | NP_001185909.1 | 7 | 625 | |||
NM_022733.2 | NP_073570.1 | 7 | 1052 | |||
XM_011541960.1 | XP_011540262.1 | 8 | 1059 | |||
XM_017002093.1 | XP_016857582.1 | 4 | 751 | |||
GenBank mRNA | AK294675.1 | - | 2 | 334 | ||
AK298702.1 | - | 7 | 590 | |||
AK303975.1 | - | 7 | 598 | |||
AK308280.1 | - | 7 | 695 | |||
AK313105.1 | - | 7 | 999 | |||
AK316218.1 | - | 7 | 625 | |||
AL133206.1 | - | 7 | 566 | |||
AL137764.1 | - | 7 | 579 | |||
BC014512.1 | - | 5 | 391 | |||
BC021133.1 | - | 7 | 702 | 4855320 |