Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134774.1 | NP_001128246.1 | 4, 5 | 696 | ||
NM_001134775.1 | NP_001128247.1 | 5, 6 | 990 | |||
NM_001134776.1 | NP_001128248.1 | 5, 6 | 869 | |||
NM_001318734.1 | NP_001305663.1 | 5, 6 | 927 | |||
NM_022822.2 | NP_073733.1 | 5, 6 | 964 | |||
XM_005274183.1 | XP_005274240.1 | 5, 6 | 906 | |||
XM_005274184.1 | XP_005274241.1 | 5, 6 | 927 | |||
GenBank mRNA | AK022449.1 | - | 5, 6 | 856 | ||
AK022907.1 | - | 5, 6 | 909 | |||
AK094593.1 | - | 3, 4 | 566 | |||
AK126489.1 | - | 3, 4 | 1348 | |||
AK315725.1 | - | 5, 6 | 859 | |||
AL136864.1 | - | 5, 6 | 890 | |||
BC034373.1 | - | 5, 6 | 852 | 4342879 | ||
BE882786.1 | - | 4, 5 | 591 |