Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST8SIA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304450.1 | NP_001291379.1 | 4 | 1068 | ||
NM_003034.3 | NP_003025.1 | 5 | 1178 | |||
GenBank mRNA | AK290941.1 | - | 5 | 819 | ||
AK297310.1 | - | 4 | 551 | |||
AK315340.1 | - | 5 | 1153 | |||
AY569975.1 | - | 4 | 592 | |||
BC117285.1 | - | 5 | 1097 | 40125836 | ||
BC126162.1 | - | 5 | 1097 | 8991878 | ||
D26360.1 | - | 5 | 799 | |||
EU041716.1 | - | 4 | 1068 | |||
L32867.1 | - | 5 | 763 | |||
L43494.1 | - | 5 | 707 | |||
X77922.1 | - | 5 | 1178 |