Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INTS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324475.1 | NP_001311404.1 | 12 | 1400 | ||
NM_023015.4 | NP_075391.3 | 11 | 1741 | |||
XM_005245459.2 | XP_005245516.1 | 11 | 1758 | |||
XM_005245461.2 | XP_005245518.1 | 4 | 341 | |||
XM_006711490.3 | XP_006711553.1 | 12 | 1211 | |||
XM_011509906.2 | XP_011508208.1 | 11 | 1123 | |||
GenBank mRNA | AK074979.1 | - | 4 | 439 | ||
AK290025.1 | - | 12 | 1394 | |||
AK304874.1 | - | 5 | 830 | |||
AL080172.1 | - | 5 | 550 | |||
AL832133.1 | - | 10 | 2776 | |||
BC105092.1 | - | 11 | 1204 | 8144095 | ||
BC105094.1 | - | 11 | 1204 | 8144097 | ||
BC116458.1 | - | 11 | 1736 | 5752962 | ||
BX640786.1 | - | 11 | 1727 | |||
BX640950.1 | - | 11 | 1742 | |||
CR627233.1 | - | 8 | 5038 | |||
CR749212.1 | - | 9 | 3268 | |||
CR749376.1 | - | 10 | 3393 |