Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006516.2 | NP_006507.2 | 5, 6 | 1200 | ||
GenBank mRNA | AB208987.1 | - | 5, 6 | 920 | ||
AF070544.1 | - | 2, 3 | 228 | |||
AK122999.1 | - | 3, 4 | 460 | |||
AK292791.1 | - | 5, 6 | 886 | |||
AK293306.1 | - | 5, 6 | 883 | |||
AK296736.1 | - | 6, 7 | 718 | |||
AK312403.1 | - | 5, 6 | 892 | |||
BC118590.1 | - | 5, 6 | 821 | 40085221 | ||
BC121804.1 | - | 5, 6 | 821 | 40085220 | ||
BG682043.1 | - | 1, 2 | 43 | |||
K03195.1 | - | 5, 6 | 854 |