Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201.2 | NP_001192.2 | 2 | 820 | ||
XM_006714291.3 | XP_006714354.1 | 2 | 1007 | |||
GenBank mRNA | BC096269.2 | - | 2 | 771 | 40006302 | |
BC096270.2 | - | 2 | 771 | 40006303 | ||
BC096271.2 | - | 2 | 771 | 40006304 | ||
BC117514.1 | - | 2 | 771 | 40006306 | ||
M22491.1 | - | 2 | 820 |