Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM228A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040710.2 | NP_001035800.1 | 6 | 615 | ||
GenBank mRNA | BC050462.1 | - | 6 | 555 | 5744344 |