Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003090.3 | NP_003081.2 | 1 | 116 | ||
NR_135506.1 | 1 | 116 | ||||
NR_135507.1 | 1 | 116 | ||||
NR_135508.1 | 1 | 116 | ||||
GenBank mRNA | AJ130971.1 | - | 1 | 17 | ||
AJ130972.1 | - | 1 | 17 | |||
AK223070.1 | - | 1 | 73 | |||
AK223110.1 | - | 1 | 38 | |||
AK290078.1 | - | 1 | 72 | |||
AK312200.1 | - | 1 | 47 | |||
BC022816.1 | - | 1 | 69 | 4904205 | ||
BC067846.1 | - | 1 | 85 | 30330402 | ||
BC071717.1 | - | 1 | 24 | 4514975 | ||
X13482.1 | - | 1 | 68 |