Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137601.2 | NP_001131073.1 | 1 | 1239 | ||
XM_017020911.1 | XP_016876400.1 | 2 | 1343 | |||
GenBank mRNA | AK125003.1 | - | 1 | 355 | ||
AX721091.1 | - | 2 | 1344 | |||
BC157833.1 | - | 1 | 1249 | 9057052 | ||
BC171822.1 | - | 1 | 1249 | 9054476 |