Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SQLE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003129.3 | NP_003120.2 | 6 | 1921 | ||
XM_011517246.2 | XP_011515548.1 | 6 | 1019 | |||
GenBank mRNA | AF098865.1 | - | 6 | 1150 | ||
AK225085.1 | - | 6 | 1211 | |||
AK295302.1 | - | 6 | 821 | |||
AK301792.1 | - | 6 | 813 | |||
AK313384.1 | - | 6 | 1039 | |||
BC017033.1 | - | 6 | 1153 | 3846346 | ||
D78129.1 | - | 4 | 439 |