Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TADA2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166105.2 | NP_001159577.2 | 3 | 233 | ||
NM_001291918.1 | NP_001278847.1 | 3 | 233 | |||
NM_001488.4 | NP_001479.4 | 3 | 441 | |||
NM_133439.3 | NP_597683.3 | 3 | 441 | |||
XM_006722044.3 | XP_006722107.1 | 3 | 233 | |||
XM_017024981.1 | XP_016880470.1 | 3 | 229 | |||
XM_017024982.1 | XP_016880471.1 | 3 | 233 | |||
XM_017024983.1 | XP_016880472.1 | 3 | 233 | |||
XM_017024984.1 | XP_016880473.1 | 3 | 242 | |||
XM_017024985.1 | XP_016880474.1 | 3 | 242 | |||
XM_017024986.1 | XP_016880475.1 | 3 | 233 | |||
GenBank mRNA | AF064094.1 | - | 2 | 180 | ||
AK022767.1 | - | 3 | 444 | |||
AK096371.1 | - | 3 | 91 | |||
AK225918.1 | - | 3 | 230 | |||
AL526501.3 | - | 3 | 211 | |||
BC001172.1 | - | 2 | 180 | 3355742 | ||
BC011753.2 | - | 2 | 180 | 3533186 | ||
CD101741.1 | - | 3 | 268 | |||
DA244842.1 | - | 3 | 233 | |||
KU178344.1 | - | 2 | 72 | |||
KU178345.1 | - | 2 | 72 |