Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003223.2 | NP_003214.1 | 1, 2 | 338 | ||
GenBank mRNA | AK309301.1 | - | 1, 2 | 405 | ||
BC010576.1 | - | 1, 2 | 227 | 4181538 | ||
BU159415.1 | - | 1, 2 | 291 | |||
CR541726.1 | - | 1, 2 | 84 | |||
DA449763.1 | - | 1, 2 | 405 | |||
S73885.1 | - | 1, 2 | 342 | |||
X57435.1 | - | 1, 2 | 33 |