Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003225.2 | NP_003216.1 | 1, 2 | 122 | ||
GenBank mRNA | BC032811.1 | - | 1, 2 | 106 | 5186922 | |
BQ083523.1 | - | 1, 2 | 181 | |||
BU539534.1 | - | 383 | ||||
M12075.1 | - | 1, 2 | 81 | |||
X00474.1 | - | 1, 2 | 122 | |||
X52003.1 | - | 1, 2 | 122 |