Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144981.2 | NP_001138453.1 | 1, 2 | 230 | ||
NM_001198908.1 | NP_001185837.1 | 1, 2 | 230 | |||
GenBank mRNA | AK131058.1 | - | 1, 2 | 181 | ||
BC146975.1 | - | 1, 2 | 124 | 9056903 | ||
DB454585.1 | - | 1, 2 | 231 | |||
DN999189.1 | - | 1, 2 | 187 |