Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCDC2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287444.1 | NP_001274373.1 | 1, 2 | 449 | ||
XM_006711894.3 | XP_006711957.1 | 1, 2 | 443 | |||
XM_011510393.2 | XP_011508695.1 | 1, 2 | 443 | |||
XM_011510394.2 | XP_011508696.1 | 1, 2 | 443 | |||
XM_017004836.1 | XP_016860325.1 | 1, 2 | 443 | |||
XM_017004837.1 | XP_016860326.1 | 1, 2 | 443 | |||
XM_017004838.1 | XP_016860327.1 | 1, 2 | 443 | |||
GenBank mRNA | DW010024.1 | - | 1, 2 | 144 | ||
HY010234.1 | - | 1, 2 | 449 |