Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUFT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126337.1 | NP_001119809.1 | 7 | 620 | ||
NM_001301317.1 | NP_001288246.1 | 9 | 830 | |||
NM_020127.2 | NP_064512.1 | 8 | 695 | |||
XM_011509961.2 | XP_011508263.1 | 6 | 529 | |||
XM_017002223.1 | XP_016857712.1 | 8 | 771 | |||
XM_017002224.1 | XP_016857713.1 | 7 | 892 | |||
GenBank mRNA | AF254260.1 | - | 8 | 695 | ||
AK075001.1 | - | 7 | 617 | |||
AK223161.1 | - | 8 | 691 | |||
AK298066.1 | - | 9 | 830 | |||
AK299925.1 | - | 7 | 608 | |||
AK315414.1 | - | 8 | 691 | |||
AL136917.1 | - | 8 | 683 | |||
AY358610.1 | - | 7 | 614 | |||
BC002933.1 | - | 7 | 588 | 3956873 | ||
BC008301.1 | - | 8 | 675 | 3506004 |