Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015410.1 | 2 | 321 | |||
GenBank mRNA | AK026189.1 | - | 2 | 321 | ||
AK300802.1 | - | 2 | 350 | |||
AK310212.1 | - | 3 | 441 |