Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267723.1 | NP_001254652.1 | 4, 5 | 439 | ||
XM_005259191.3 | XP_005259248.1 | 4, 5 | 2544 | |||
XM_005259192.4 | XP_005259249.1 | 4, 5 | 1185 | |||
XM_011527257.2 | XP_011525559.1 | 4, 5 | 468 | |||
GenBank mRNA | BI819372.1 | - | 3, 4 | 339 | ||
DR007040.1 | - | 4, 5 | 459 | |||
GQ375050.1 | - | 4, 5 | 412 |