Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUFY4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198483.3 | NP_940885.2 | 2 | 366 | ||
NR_034176.1 | 2 | 283 | ||||
XM_011511014.2 | XP_011509316.1 | 1 | 389 | |||
XM_017003895.1 | XP_016859384.1 | 1 | 389 | |||
XM_017003896.1 | XP_016859385.1 | 1 | 389 | |||
XM_017003897.1 | XP_016859386.1 | 1 | 389 | |||
GenBank mRNA | AK128393.1 | - | 2 | 229 | ||
AK131246.1 | - | 1 | 258 | |||
BX641664.1 | - | 2 | 284 | |||
DA958158.1 | - | 2 | 366 |