Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WIPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077269.1 | NP_001070737.1 | 3 | 324 | ||
NM_003387.4 | NP_003378.3 | 3 | 260 | |||
XM_011511780.2 | XP_011510082.1 | 3 | 446 | |||
GenBank mRNA | AF031588.1 | - | 3 | 268 | ||
AK301271.1 | - | 3 | 265 | |||
AK304194.1 | - | 3 | 277 | |||
AL529597.3 | - | 3 | 292 | |||
BC002914.2 | - | 3 | 352 | 3954968 | ||
BC110288.1 | - | 3 | 296 | 5184105 | ||
BQ441937.1 | - | 1 | 61 | |||
BX640870.1 | - | 3 | 324 | |||
X86019.1 | - | 3 | 364 |