Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291880.1 | NP_001278809.1 | 2 | 1138 | ||
NM_004185.4 | NP_004176.2 | 3 | 1209 | |||
NM_024494.2 | NP_078613.1 | 2 | 784 | |||
GenBank mRNA | AB045116.1 | - | 3 | 332 | ||
AB045117.1 | - | 2 | 288 | |||
AK127449.1 | - | 2 | 1138 | |||
AK312696.1 | - | 2 | 536 | |||
BC141825.1 | - | 2 | 784 | 40148248 |