Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278284.1 | NP_001265213.1 | 6 | 1722 | ||
NM_001278287.1 | NP_001265216.1 | 6 | 1638 | |||
NM_001278290.1 | NP_001265219.1 | 5 | 1483 | |||
NM_001278291.1 | NP_001265220.1 | 4 | 1352 | |||
NM_001278292.1 | NP_001265221.1 | 5 | 1473 | |||
NM_001318135.1 | NP_001305064.1 | 6 | 1600 | |||
NM_001318136.1 | NP_001305065.1 | 3 | 1632 | |||
NM_032924.4 | NP_116313.3 | 6 | 1604 | |||
XM_006716111.1 | XP_006716174.1 | 6 | 1658 | |||
XM_017012582.1 | XP_016868071.1 | 6 | 1672 | |||
XM_017012583.1 | XP_016868072.1 | 4 | 1821 | |||
GenBank mRNA | AK091984.1 | - | 3 | 1604 | ||
AK315479.1 | - | 4 | 1318 | |||
BC011887.2 | - | 6 | 1528 | 3831842 | ||
BC013603.1 | - | 6 | 1547 | 3865426 | ||
BC025265.1 | - | 6 | 1537 | |||
BT006918.1 | - | 3 | 1169 | |||
BX647182.1 | - | 5 | 2245 | |||
X07290.1 | - | 1 | 526 |