Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021916.3 | NP_068735.1 | 2 | 1276 | ||
GenBank mRNA | AK122721.1 | - | 1 | 843 | ||
BC040161.1 | - | 2 | 1276 | 5551072 | ||
CR456615.1 | - | 2 | 949 |