Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF75D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007131.4 | NP_009062.2 | 4, 5 | 3322 | ||
XM_005262469.1 | XP_005262526.1 | 4, 5 | 1851 | |||
XM_011531391.1 | XP_011529693.1 | 5, 6 | 2038 | |||
XM_011531396.2 | XP_011529698.1 | 3, 4 | 2815 | |||
GenBank mRNA | AK092347.1 | - | 1, 2 | 211 | ||
AK290215.1 | - | 3, 4 | 1026 | |||
AL831984.2 | - | 4, 5 | 3267 | |||
AL832801.1 | - | 3, 4 | 809 | |||
BC109100.2 | - | 3, 4 | 841 | 40034784 |