Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CACNA1C SIRNA Assays ›
Gene Symbol:
CACNA1C
Gene Name:
calcium voltage-gated channel subunit alpha1 C
Gene Aliases:
CACH2, CACN2, CACNL1A1, CCHL1A1, CaV1.2, LQT8, TS
Chromosome Location:
Chr.12: 1969677 - 2697949 on Build GRCh38
UniGene ID:
Hs.118262
Species:
Human
siRNA ID
HSS187850
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CACNA1C
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000719.6
NP_000710.5
26
3602
NM_001129827.1
NP_001123299.1
27
3662
NM_001129829.1
NP_001123301.1
26
3602
NM_001129830.2
NP_001123302.2
26
3567
NM_001129831.1
NP_001123303.1
26
3602
NM_001129832.1
NP_001123304.1
27
3662
NM_001129833.1
NP_001123305.1
26
3602
NM_001129834.1
NP_001123306.1
26
3602
NM_001129835.1
NP_001123307.1
26
3602
NM_001129836.1
NP_001123308.1
26
3602
NM_001129837.1
NP_001123309.1
26
3602
NM_001129838.1
NP_001123310.1
26
3602
NM_001129839.1
NP_001123311.1
26
3602
NM_001129840.1
NP_001123312.1
26
3602
NM_001129841.1
NP_001123313.1
26
3602
NM_001129842.1
NP_001123314.1
26
3602
NM_001129843.1
NP_001123315.1
26
3602
NM_001129844.1
NP_001123316.1
26
3593
NM_001129846.1
NP_001123318.1
26
3602
NM_001167623.1
NP_001161095.1
26
3602
NM_001167624.2
NP_001161096.2
26
3567
NM_001167625.1
NP_001161097.1
26
3602
NM_199460.3
NP_955630.3
27
3627
XM_006719017.2
XP_006719080.1
26
3656
XM_011521020.2
XP_011519322.1
27
3731
XM_011521023.2
XP_011519325.1
27
3916
XM_017019926.1
XP_016875415.1
29
5038
XM_017019927.1
XP_016875416.1
29
5038
XM_017019928.1
XP_016875417.1
29
5038
XM_017019929.1
XP_016875418.1
29
5038
XM_017019930.1
XP_016875419.1
29
5038
XM_017019931.1
XP_016875420.1
29
5038
XM_017019932.1
XP_016875421.1
27
4843
XM_017019933.1
XP_016875422.1
27
4843
XM_017019934.1
XP_016875423.1
28
4918
XM_017019935.1
XP_016875424.1
28
4918
XM_017019936.1
XP_016875425.1
27
4843
XM_017019937.1
XP_016875426.1
27
4843
XM_017019938.1
XP_016875427.1
27
4843
XM_017019939.1
XP_016875428.1
27
4843
XM_017019940.1
XP_016875429.1
27
4843
XM_017019941.1
XP_016875430.1
27
4843
XM_017019942.1
XP_016875431.1
27
4843
XM_017019943.1
XP_016875432.1
27
4843
XM_017019944.1
XP_016875433.1
27
4843
XM_017019945.1
XP_016875434.1
27
4843
XM_017019946.1
XP_016875435.1
27
4843
XM_017019947.1
XP_016875436.1
27
4843
XM_017019948.1
XP_016875437.1
27
4843
XM_017019949.1
XP_016875438.1
27
4843
XM_017019950.1
XP_016875439.1
27
4843
XM_017019951.1
XP_016875440.1
27
4834
XM_017019952.1
XP_016875441.1
27
4843
XM_017019953.1
XP_016875442.1
26
3656
XM_017019954.1
XP_016875443.1
26
3656
XM_017019955.1
XP_016875444.1
28
4918
GenBank mRNA
AB209016.1
-
24
3037
AF465484.1
-
26
3554
AJ224873.1
-
26
3289
AY830711.1
-
26
3289
AY830712.1
-
26
3289
AY830713.1
-
26
3289
BC146846.1
-
26
3408
9056774
L29529.1
-
25
3221
L29534.1
-
25
3215
L29536.1
-
25
3215
Z26295.1
-
1
16
Z34809.1
-
26
3289
Z34810.1
-
26
3289
Z34811.1
-
26
3289
Z34812.1
-
26
3289
Z34813.1
-
26
3289
Z34814.1
-
26
3289
Z34815.1
-
26
3289
Z34816.1
-
26
3289
Z34817.1
-
26
3289
Z34818.1
-
27
3349
Z34819.1
-
26
3289
Z34820.1
-
26
3289
Z34821.1
-
27
3349
Z34822.1
-
26
3289
Z74996.1
-
26
3289
Back To Top
More Information
Panther Classification:
Molecular Function -
voltage-gated ion channel
Gene Ontology Categories:
Function(s)
Process(es)
immune system development
positive regulation of cytosolic calcium ion concentration
heart development
regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
embryonic forelimb morphogenesis
calcium-mediated signaling using extracellular calcium source
camera-type eye development
regulation of insulin secretion
calcium ion transport into cytosol
cardiac conduction
calcium ion transmembrane transport via high voltage-gated calcium channel
calcium ion transmembrane transport
cardiac muscle cell action potential involved in contraction
membrane depolarization during cardiac muscle cell action potential
membrane depolarization during AV node cell action potential
cell communication by electrical coupling involved in cardiac conduction
regulation of heart rate by cardiac conduction
regulation of ventricular cardiac muscle cell action potential
membrane depolarization during atrial cardiac muscle cell action potential
voltage-gated calcium channel activity
protein binding
calmodulin binding
high voltage-gated calcium channel activity
metal ion binding
alpha-actinin binding
voltage-gated calcium channel activity involved in cardiac muscle cell action potential
voltage-gated calcium channel activity involved in AV node cell action potential
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0