Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EVI5 SIRNA Assays ›
Gene Symbol:
EVI5
Gene Name:
ecotropic viral integration site 5
Gene Aliases:
EVI-5, NB4S
Chromosome Location:
Chr.1: 92508696 - 92792404 on Build GRCh38
UniGene ID:
Hs.594434
Species:
Human
siRNA ID
HSS187883
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EVI5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001308248.1
NP_001295177.1
16, 17
1959
NM_005665.5
NP_005656.4
15, 16
1926
XM_017002269.1
XP_016857758.1
18, 19
2185
XM_017002270.1
XP_016857759.1
18, 19
2496
XM_017002271.1
XP_016857760.1
17, 18
2104
XM_017002272.1
XP_016857761.1
18, 19
2185
XM_017002273.1
XP_016857762.1
18, 19
2411
XM_017002274.1
XP_016857763.1
18, 19
2160
XM_017002275.1
XP_016857764.1
18, 19
2148
XM_017002276.1
XP_016857765.1
17, 18
2328
XM_017002277.1
XP_016857766.1
16, 17
1957
XM_017002278.1
XP_016857767.1
17, 18
2038
XM_017002279.1
XP_016857768.1
17, 18
2097
XM_017002280.1
XP_016857769.1
17, 18
2328
XM_017002281.1
XP_016857770.1
16, 17
2259
XM_017002282.1
XP_016857771.1
18, 19
2185
XM_017002283.1
XP_016857772.1
17, 18
2104
XM_017002284.1
XP_016857773.1
16, 17
2180
XM_017002285.1
XP_016857774.1
17, 18
2327
XM_017002286.1
XP_016857775.1
16, 17
2098
XM_017002287.1
XP_016857776.1
17, 18
2179
XM_017002288.1
XP_016857777.1
15, 16
1721
GenBank mRNA
AB209513.1
-
11, 12
1105
AB449875.1
-
16, 17
1949
AF008915.1
-
15, 16
1927
Back To Top
More Information
Panther Classification:
Molecular Function -
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
intracellular protein transport
cell cycle
multicellular organism development
cell proliferation
regulation of vesicle fusion
retrograde transport, endosome to Golgi
cell division
activation of GTPase activity
regulation of cilium assembly
GTPase activator activity
protein binding
Rab GTPase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0