Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COLEC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001255982.1 | NP_001242911.1 | 6 | 926 | ||
NM_001255983.1 | NP_001242912.1 | 6 | 926 | |||
NM_001255984.1 | NP_001242913.1 | 5 | 854 | |||
NM_001255985.1 | NP_001242914.1 | 8 | 888 | |||
NM_001255986.1 | NP_001242915.1 | 6 | 697 | |||
NM_001255987.1 | NP_001242916.1 | 5 | 625 | |||
NM_001255988.1 | NP_001242917.1 | 5 | 625 | |||
NM_001255989.1 | NP_001242918.1 | 4 | 553 | |||
NM_024027.4 | NP_076932.1 | 7 | 998 | |||
NM_199235.2 | NP_954705.1 | 8 | 1077 | |||
NR_045659.1 | 8 | 1191 | ||||
XM_005263853.4 | XP_005263910.1 | 6 | 620 | |||
XM_006711897.3 | XP_006711960.1 | 6 | 620 | |||
GenBank mRNA | AB119525.1 | - | 7 | 884 | ||
AB119650.1 | - | 6 | 812 | |||
AB119651.1 | - | 6 | 812 | |||
AB119652.1 | - | 5 | 740 | |||
AB119684.1 | - | 6 | 697 | |||
AB119685.1 | - | 5 | 625 | |||
AB119686.1 | - | 5 | 625 | |||
AB119687.1 | - | 4 | 553 | |||
AK303824.1 | - | 8 | 888 | |||
AK313840.1 | - | 7 | 1000 | |||
AY358439.1 | - | 7 | 701 | |||
BC000078.2 | - | 7 | 708 | 3507377 | ||
BC009951.2 | - | 8 | 860 | 4125795 | ||
CR936641.1 | - | 5 | 489 |