Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LBHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024099.3 | NP_077004.2 | 6 | 962 | ||
GenBank mRNA | AK055736.1 | - | 4 | 2553 | ||
BC001434.2 | - | 6 | 962 | 3138904 | ||
BI831485.1 | - | 252 |
Gene Symbol: C11orf98 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286086.1 | NP_001273015.1 | 3 | 294 | ||
NR_104414.1 | 2 | 451 | ||||
GenBank mRNA | BF207356.1 | - | 3 | 279 | ||
BG563566.1 | - | 3 | 283 | |||
BG681685.1 | - | 2 | 395 | |||
BQ231052.1 | - | 3 | 276 | |||
BX107288.1 | - | 3 | 266 | |||
CN256679.1 | - | 3 | 295 |