Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MFSD11 SIRNA Assays ›
Gene Symbol:
MFSD11
Gene Name:
major facilitator superfamily domain containing 11
Gene Aliases:
ET
Chromosome Location:
Chr.17: 76736565 - 76803805 on Build GRCh38
UniGene ID:
Hs.732019
Species:
Human
siRNA ID
HSS188009
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MFSD11
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242532.1
NP_001229461.1
7
1413
NM_001242533.1
NP_001229462.1
8
1297
NM_001242534.1
NP_001229463.1
8
1159
NM_001242535.1
NP_001229464.1
8
1108
NM_001242536.1
NP_001229465.1
6
1141
NM_001242537.1
NP_001229466.1
6
952
NM_024311.3
NP_077287.1
8
1015
XM_011525236.2
XP_011523538.1
7
677
XM_011525237.2
XP_011523539.2
8
1120
XM_011525238.2
XP_011523540.2
8
720
XM_011525239.2
XP_011523541.2
8
690
XM_011525240.2
XP_011523542.2
8
716
XM_011525241.2
XP_011523543.2
8
726
XM_011525242.1
XP_011523544.1
7
677
XM_011525244.1
XP_011523546.1
7
677
XM_011525247.2
XP_011523549.2
8
1160
XM_017025065.1
XP_016880554.1
8
748
XM_017025066.1
XP_016880555.1
8
690
XM_017025067.1
XP_016880556.1
8
748
XM_017025068.1
XP_016880557.1
8
1160
XM_017025069.1
XP_016880558.1
6
848
XM_017025070.1
XP_016880559.1
6
848
GenBank mRNA
AF015184.1
-
8
727
AF015185.1
-
6
758
AF015186.1
-
8
871
AK000233.1
-
7
795
AK025849.1
-
7
550
AK308058.1
-
6
596
BC002753.2
-
8
815
3629029
BC062563.1
-
8
908
5140496
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
molecular_function
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0