Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122835.2 | NP_001116307.2 | 9 | 1037 | ||
NM_024525.4 | NP_078801.3 | 11 | 1196 | |||
XM_005273260.1 | XP_005273317.1 | 11 | 1196 | |||
XM_005273261.2 | XP_005273318.1 | 10 | 1103 | |||
XM_005273262.1 | XP_005273319.1 | 9 | 1037 | |||
XM_005273264.2 | XP_005273321.1 | 9 | 968 | |||
XM_006711814.2 | XP_006711877.1 | 7 | 809 | |||
XM_006711815.2 | XP_006711878.1 | 4 | 2203 | |||
XM_011544275.2 | XP_011542577.1 | 8 | 875 | |||
XM_011544276.2 | XP_011542578.1 | 4 | 439 | |||
XM_017002321.1 | XP_016857810.1 | 10 | 1103 | |||
XM_017002322.1 | XP_016857811.1 | 9 | 968 | |||
XM_017002323.1 | XP_016857812.1 | 7 | 771 | |||
XM_017002324.1 | XP_016857813.1 | 7 | 809 | |||
XM_017002325.1 | XP_016857814.1 | 11 | 4401 | |||
XM_017002326.1 | XP_016857815.1 | 6 | 3781 | |||
GenBank mRNA | AK074365.1 | - | 2 | 103 | ||
AK075386.1 | - | 11 | 1162 | |||
AK075561.1 | - | 9 | 1016 | |||
AK309725.1 | - | 7 | 855 | |||
AL832286.1 | - | 11 | 931 | |||
AL834453.1 | - | 2 | 165 | |||
BC042683.1 | - | 2 | 83 |