Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADIPOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024551.2 | NP_078827.2 | 4 | 644 | ||
XM_005253789.1 | XP_005253846.1 | 4 | 766 | |||
XM_006719018.2 | XP_006719081.1 | 5 | 763 | |||
XM_011521024.2 | XP_011519326.1 | 5 | 774 | |||
GenBank mRNA | AK025085.1 | - | 1 | 172 | ||
AK127196.1 | - | 5 | 699 | |||
AK128511.1 | - | 4 | 698 | |||
AY424280.1 | - | 3 | 523 | |||
BC051858.2 | - | 4 | 644 | 6527208 |