Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISOC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136201.1 | NP_001129673.1 | 2 | 297 | ||
NM_001136202.1 | NP_001129674.1 | 2 | 297 | |||
NM_024710.2 | NP_078986.1 | 2 | 297 | |||
GenBank mRNA | AK027122.1 | - | 2 | 260 | ||
AK131324.1 | - | 2 | 958 | |||
AK311540.1 | - | 247 | ||||
BC017344.2 | - | 2 | 184 | 4893552 | ||
BP226043.1 | - | 2 | 298 | |||
CR457377.1 | - | 1 | 123 |