Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISOC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136201.1 | NP_001129673.1 | 3 | 506 | ||
NM_024710.2 | NP_078986.1 | 3 | 506 | |||
GenBank mRNA | AK027122.1 | - | 3 | 469 | ||
AK311540.1 | - | 1146 | ||||
BC017344.2 | - | 3 | 393 | 4893552 | ||
BP226043.1 | - | 3 | 507 | |||
CR457377.1 | - | 2 | 332 |