Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNDC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024715.3 | NP_078991.3 | 5 | 1685 | ||
GenBank mRNA | AK026278.1 | - | 6 | 1088 | ||
AK056512.1 | - | 5 | 1720 | |||
AK092425.1 | - | 3 | 1750 | |||
AK225454.1 | - | 6 | 1088 | |||
AK225462.1 | - | 6 | 1088 | |||
AK299578.1 | - | 5 | 978 | |||
AK307529.1 | - | 4 | 596 | |||
AY358649.1 | - | 5 | 1113 | |||
BC001615.1 | - | 5 | 1172 | 3544923 | ||
BC032568.1 | - | 5 | 1086 | 5225890 | ||
BX538131.1 | - | 5 | 1012 |