Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105247.1 | NP_001098717.1 | 3 | 1636 | ||
NM_001288767.1 | NP_001275696.1 | 5 | 1516 | |||
NM_001301820.1 | NP_001288749.1 | 4 | 1651 | |||
NM_024742.2 | NP_079018.1 | 3 | 1636 | |||
XM_006721091.2 | XP_006721154.1 | 4 | 1769 | |||
GenBank mRNA | AK023125.1 | - | 3 | 687 | ||
AK024430.1 | - | 3 | 1129 | |||
AK294897.1 | - | 4 | 1558 | |||
AK295790.1 | - | 5 | 1516 | |||
AY217348.1 | - | 3 | 1636 | |||
BC014945.1 | - | 4 | 1041 | 4860377 |