Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMKMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024766.4 | NP_079042.1 | 2 | 364 | ||
XM_011533111.2 | XP_011531413.1 | 2 | 330 | |||
XM_017004970.1 | XP_016860459.1 | 2 | 554 | |||
XM_017004971.1 | XP_016860460.1 | 2 | 554 | |||
XM_017004972.1 | XP_016860461.1 | 2 | 554 | |||
XM_017004973.1 | XP_016860462.1 | 2 | 554 | |||
XM_017004974.1 | XP_016860463.1 | 2 | 554 | |||
XM_017004975.1 | XP_016860464.1 | 2 | 554 | |||
XM_017004976.1 | XP_016860465.1 | 2 | 554 | |||
XM_017004977.1 | XP_016860466.1 | 2 | 554 | |||
XM_017004978.1 | XP_016860467.1 | 2 | 554 | |||
XM_017004979.1 | XP_016860468.1 | 2 | 554 | |||
XM_017004980.1 | XP_016860469.1 | 2 | 554 | |||
XM_017004981.1 | XP_016860470.1 | 2 | 554 | |||
GenBank mRNA | AK027104.1 | - | 2 | 304 | ||
BC029359.1 | - | 2 | 316 | |||
BC053733.1 | - | 2 | 291 | 5199179 | ||
DA760277.1 | - | 2 | 366 | |||
DB461740.1 | - | 2 | 346 |