Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBLL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284291.1 | NP_001271220.1 | 6 | 1210 | ||
NM_024814.3 | NP_079090.2 | 6 | 1213 | |||
NR_024199.2 | 7 | 1286 | ||||
XM_011516580.2 | XP_011514882.1 | 7 | 1483 | |||
XM_017012643.1 | XP_016868132.1 | 6 | 1014 | |||
XM_017012644.1 | XP_016868133.1 | 6 | 1404 | |||
XM_017012645.1 | XP_016868134.1 | 7 | 1338 | |||
GenBank mRNA | AK026762.1 | - | 6 | 769 | ||
AK293352.1 | - | 6 | 1042 | |||
BC027460.2 | - | 5 | 738 | |||
BC130529.1 | - | 6 | 765 | 40146560 | ||
BC130531.1 | - | 6 | 765 | 40146562 | ||
BC144176.1 | - | 6 | 762 | 9052696 | ||
BX348716.2 | - | 2 | 351 |