Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABEP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024816.2 | NP_079092.2 | 2, 3 | 320 | ||
GenBank mRNA | AK026935.1 | - | 2, 3 | 320 | ||
AK092299.1 | - | 2, 3 | 861 | |||
BC027584.1 | - | 2, 3 | 798 | 4827815 | ||
BC046118.1 | - | 2, 3 | 314 | |||
BC058900.1 | - | 2, 3 | 323 | 5415624 |