Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCAKD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128631.2 | NP_001122103.1 | 5 | 830 | ||
NM_001288654.2 | NP_001275583.1 | 5 | 966 | |||
NM_001288655.1 | NP_001275584.1 | 5 | 878 | |||
NM_001321326.1 | NP_001308255.1 | 5 | 776 | |||
NM_024819.6 | NP_079095.3 | 5 | 963 | |||
XM_011525262.2 | XP_011523564.1 | 5 | 995 | |||
XM_017025105.1 | XP_016880594.1 | 6 | 1068 | |||
XM_017025106.1 | XP_016880595.1 | 6 | 1010 | |||
GenBank mRNA | AK290755.1 | - | 5 | 779 | ||
BC006472.1 | - | 5 | 852 | 2822306 | ||
BC006546.2 | - | 5 | 852 | 2822306 | ||
BC018132.2 | - | 4 | 724 | 3903964 | ||
BC033299.1 | - | 4 | 863 | 4153431 |