Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167575.1 | NP_001161047.1 | 2 | 710 | ||
NM_024828.3 | NP_079104.3 | 2 | 556 | |||
GenBank mRNA | AK023719.1 | - | 2 | 530 | ||
AK300739.1 | - | 2 | 324 | |||
AK301673.1 | - | 2 | 254 | |||
BC014658.1 | - | 2 | 532 | 4648932 | ||
BC071953.1 | - | 2 | 455 | 5431506 | ||
BP228742.1 | - | 2 | 556 |