Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122841.2 | NP_001116313.1 | 8 | 830 | ||
NM_024919.4 | NP_079195.3 | 8 | 980 | |||
NR_110312.1 | 12 | 1603 | ||||
XM_011536137.1 | XP_011534439.1 | 10 | 1129 | |||
XM_011536138.1 | XP_011534440.1 | 10 | 945 | |||
XM_011536140.1 | XP_011534442.1 | 9 | 984 | |||
XM_011536141.1 | XP_011534443.1 | 9 | 924 | |||
XM_011536142.2 | XP_011534444.1 | 9 | 962 | |||
XM_011536143.1 | XP_011534445.1 | 9 | 860 | |||
XM_011536145.1 | XP_011534447.1 | 9 | 899 | |||
XM_011536146.2 | XP_011534448.1 | 5 | 1752 | |||
XM_017011317.1 | XP_016866806.1 | 9 | 1060 | |||
XM_017011318.1 | XP_016866807.1 | 5 | 1718 | |||
GenBank mRNA | AK097179.1 | - | 8 | 980 | ||
AK097579.1 | - | 8 | 830 | |||
AK125963.1 | - | 12 | 1575 | |||
AK303194.1 | - | 7 | 634 | |||
AK303407.1 | - | 7 | 757 | |||
AK316478.1 | - | 7 | 634 | |||
AL133077.1 | - | 6 | 602 | |||
BC137144.1 | - | 8 | 938 | 9021141 |